Learn More
Biotin Antibody (BK-1/39), mFluor Violet 500 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-50120MFV500
Product Code. 18626099
  • £410.00 / 0.10mL
Description
Biotin Monoclonal antibody specifically detects Biotin in Human samples. It is validated for Immunohistochemistry
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Biotin
Monoclonal
mFluor Violet 500 SE
50mM Sodium Borate
Mouse
Protein A purified
RUO
Human
Purified
Immunohistochemistry
BK-1/39
Immunohistochemistry
Vitamin B4
Biotin conjugated to keyhole limpet haemocyanin
0.1 mL
Primary
Store at 4C in the dark.
IgG1