missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Biotin-Anti-His-Tag Antibody
Biotin-Anti-His-Tag Antibody
Brand: Invitrogen PV6089
Code : 50
Additional Details : Weight : 0.50000kg
Description
Biotin-labeled anti-His-tag antibodies are used to indirectly label proteins tagged with 6XHis. This can be advantageous when working with partially purified protein, or in cases in which direct labeling of the protein using biotin negatively impacts protein activity. These antibodies can be used with our Kinase Tracers and Eu-Streptavidin to conduct LanthaScreen™ Eu Kinase Binding Assays. Learn More and See Which Kinases Have Been Validated Using This AssaySpecifications
Kinase Assay, Kinase Assay | |
Biotin | |
150kDa | |
RUO, RUO | |
Primary | |
Biotin-labeled anti-His tag antibody in HEPES buffered saline. Store at -20°C. | |
Biochemical binding/kinetic | |
ABL1 G250E, STK16, ZAP70, MAP2K1 S218D S222D, MAPK8, CAMK2B, CHEK1, STK33, IGFR1, BLK, LYNA, NEK2, CHUK, MAPKAPK5, MAP3K7/MAP3K7IP1, DAPK2, HIPK3, MAP2K1, MAP2K6, PIK3CG, ABL1 E255K, BMX, MAP4K2, MAPK9, CAMK1D, CHEK2, PIM1, STK22D, CSNK1G2, EPHA3, EPHB4, LCK, YES1, MET, PRKCI, WNK3, ABL1, ABL2, FGFR3, FGFR3 K650E, KIT V654A, AURKB, CDC42BPA, CDK1, CDK8, CDK9/Cyclin K, LYNB, CDC42BPB, CDK2, CDK7, PDGFRB, MET M1250T, RPS6KA1, LIMK1, MAP2K2, PIK3CA/PIK3R1, MAP2K6 S207E T211E, SRC, ABL1 Y253F, KIT T670I, RPS6KA2, PAK7, IRAK4, CSK |
0.5 mg⁄ml | |
Biotin-Anti-His-Tag Antibody, 25μg, 0.5mg/mL Concentration, HIT2 Clone, 150kDa Molecular Weight, Biotin Conjugate, Used For Biochemical Kinase Assays, Drug Discovery and Development, Kinase Biology, LanthaScreen™ kinase binding assays | |
25 μg | |
Kinase Biology | |
1 Each | |
Frozen | |
Dry Ice |
For Research Use Only. Not for use in diagnostic procedures.