Learn More
Agilent BioTek Microplate Washer Complete Dispense/Waste Systems
Various systems available for use with microplate washers
Brand: Agilent BioTek 7100746S
This item is not returnable.
View return policy
Description
- For use with microplate washers
Specifications
High flow vacuum pump assembly, vacuum set with level sense assembly, 4l bottles | |
115/230 V | |
Complete Dispense/Waste System | |
EL406 Microplate Washer, 405 Touch Microplate Washer, ELx 405 Microplate Washer, 405 LS Microplate Washer |