missing translation for 'onlineSavingsMsg'
Learn More

Waters Corporation BioSuite DEAE Anion-Exchange Column

Product Code. p-7236703
Encompass
Click to view available options
Length (Metric):
75 mm
150 mm
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 13137849

Brand: Waters Corporation 186002180

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Waters BioSuite ion-exchange column offerings include strong and weak, cation (CXC) and anion-exchangers (AXC) bonded to a pH stable, methacrylic ester-based polymeric resin.

  • The availability of four separation chemistries provides chromatographers with the flexibility required to develop methods that separate proteins and / or peptides based upon minor charge differences.
  • Nonporous (NP) and porous IEX columns are also available. Superior chromatographic resolution is possible using the nonporous IEX offerings however, greater binding capacity is obtained with the porous selections.
  • In addition, selected BioSuite ion-exchange columns are available in PEEK hardware as well as in 21.5 mm preparative column sizes.

Specifications

Diameter (Metric) 7.5 mm
Length (Metric) 75 mm
Particle Shape Spherical
Particle Size 10 μm
Pore Size 1000 Å
Quantity 1/Ea.
USP Type L23
Type Column
For Use With (Application) Protein
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.