missing translation for 'onlineSavingsMsg'
Learn More

Agilent BioTek BioStack Automated Microplate Stackers
GREENER_CHOICE

Product Code. 17287909 Shop All Agilent BioTek Products
Change view
Click to view available options
Product Type:
BioStack 3 Microplate Stacker
BioStack 4 Microplate Stacker
BioStack Microplate Stacker
BioStack NEO Microplate Stacker
Unit Size:
Each
Product Code. Product Type unitSize
17287909 BioStack NEO Microplate Stacker Each
17257909 BioStack Microplate Stacker Each
17277909 BioStack 4 Microplate Stacker Each
17267909 BioStack 3 Microplate Stacker Each
4 options
This item is not returnable. View return policy

Product Code. 17287909

Brand: Agilent BioTek BSTCKN2SN

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

Includes

One instrument integration kit, communication cable, operator manual, microplate pedestal for storage stack loading and unloading, maintenance kit.

BioTek BioStack microplate stacker is a compact and versatile microplate stacker, compatible with Agilent BioTek washers, dispensers, plate readers, and imaging systems.

BioTek BioStack microplate stacker is a compact and versatile microplate stacker, compatible with Agilent BioTek washers, dispensers, plate readers, and imaging systems. The rapid plate exchange speeds deliver increased throughput and enhanced efficiency and greater productivity

BioStack can accommodate assay workflows for 24- to 1536-well plates. Microscope slides can be stacked for imaging using BioTek's custom slide holders. The BioTek BioStack 4 is a delidding microplate stacker, automating the handling of lidded microplates for sensitive cell-based assays, for complete walkaway automation.

Certifications

CE marked; ETL listed

TRUSTED_SUSTAINABILITY

Specifications

Hertz 50/60 Hz
Voltage 100/240 V
Operating Temperature Range (Metric) 18 to 40°C
Certifications/Compliance CE and TUV marked. RoHS compliant.
Depth (English) 20.7 in.
Depth (Metric) 52.6 cm
Holds 10-, 30- and 50-plate stacks are removable and interchangeable; 96-/384-well plates: Up to 50 plates; 1536-well plates: Up to 75 plates
Humidity 10 to 85%
Includes One instrument integration kit; Communication cable; Operator manual; Microplate pedestal for storage stack loading and unloading; Maintenance kit
Plate Type ANSI/SBS Standard and low profile 96-, 384- and 1536-well plates, Maximum plate height 14.6 mm
Software LHC v 2.12 for liquid handling instruments (optional); Gen5 2.01 for readers
Weight (English) <25 lb.
Weight (Metric) 11.3 kg
Width (English) 7.4 in.
Width (Metric) 18.8 cm
Product Type BioStack NEO Microplate Stacker
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.