missing translation for 'onlineSavingsMsg'
Learn More

Agilent BioTek BioSpa 8 Automated Incubator
GREENER_CHOICE

Product Code. 17247909 Shop All Agilent BioTek Products
Change view
Click to view available options
Includes:
1 Alignment Kit
CO2/O2 Control and Monitoring, Water Pan and Humidity Monitoring, 1 Alignment Kit
Unit Size:
Each
Product Code. Includes unitSize
17247909 CO2/O2 Control and Monitoring, Water Pan and Humidity Monitoring, 1 Alignment Kit Each
17237909 1 Alignment Kit Each
2 options
This item is not returnable. View return policy

Product Code. 17247909

Brand: Agilent BioTek BIOSPAGSN

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

The BioTek BioSpa live cell analysis system offers unique benefits for a wide variety of live cell imaging applications in up to 8 microplates or other labware. For Research Use Only. Not for use in diagnostic procedures.

The BioTek BioSpa live cell analysis system offers unique benefits for a wide variety of live cell imaging applications in up to 8 microplates or other labware. With 5 imaging modes, the system enables a wide range of microscopy applications

Capture detailed cellular images, get cell-level analyses for migration and wound healing assays, 3D spheroid formation, and many other kinetic applications. Built-in scheduling, environmental monitoring, and available liquid handling for walk away automation. Multiple users can run simultaneous processes for maximum efficiency in the lab.

Warranty

1 year

TRUSTED_SUSTAINABILITY

Specifications

Operating Temperature Range (Metric) Ambient to 45°C
Certifications/Compliance CE and TUV marked. RoHS compliant.
Depth (English) 20.1 in.
Depth (Metric) 51 cm
Height (English) 18.9 in.
Height (Metric) 48 cm
Humidity 80 to 95%
Includes CO2/O2 Control and Monitoring, Water Pan and Humidity Monitoring, 1 Alignment Kit
Software BioSpa Softwate
Width (English) 27.2 in.
Width (Metric) 69 cm
Product Type Automated Microplate Incubator
Capacity 8-plate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.