missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ BIOShield™ 720 Rotor Tube Adapters

Enhance your rotor's versatility with Thermo Scientific™ BIOShield™ 720 Tube Adapters, which support a wide assortment of tube sizes.

Brand:  Thermo Scientific™ 75003823

Code : 51

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 10154464

  • £443.00 / Pack of 4
Estimated Shipment: 14-10-2022
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Enhance your rotor's versatility with Thermo Scientific™ BIOShield™ 720 Tube Adapters, which support a wide assortment of tube sizes. These adapters are for use with the BIOShield 720 High Speed Swinging-Bucket Rotor.

  • Maximum tube capacity: 4 × 150mL
  • Maximum O.D. × L: 50 × 122mm
Specifications

Specifications

1.5/2.0mL Conical Microtube
Videos
SDS
Documents
Certificates
Special Offers

Special Offers