Learn More
AHLSTROM Biosample RUO with Perforated Circles, with Cover
Product Code. 17875092
Click to view available options
Includes:
5 Perforated Circles
Unit Size:
Pack of 100
Product Code. 17875092
Brand: AHLSTROM 8.460.0005.BN
Biospecimen microsampling made easy. BioSample, Autocollect and 5 circles cards are designed for micro-volume sampling, transport and ambient storage of biological samples.
These cards are made of white and pure absorbent fibers and contain no wet-strength additives or chemicals.
Key Areas Of Use Include:
  • HIV viral load testing
  • Infectious diseasesscreening (incl. Hepatitis B, C and Syphilis)
  • Protein-based analysis
  • Genomics
  • Human Identification
  • Forensics
  • New born screening
TRUSTED_SUSTAINABILITY
Specifications
Product Type Specimen Collection and Transport Card
Closure Included Yes
Graduated No
Material Pure Absorbent Fibers
Shape Rectangular
Sterility Non-sterile
Capacity (Metric) 70 μL/Sample Area
Certifications/Compliance ISO 9001
Includes 5 Perforated Circles