missing translation for 'onlineSavingsMsg'
Learn More

Waters Corporation BioResolve-RP-mAb Column Method Validation Kit

Product Code. p-7236819
Encompass
Change view
Click to view available options
Length (Metric):
100 mm
150 mm
Diameter (Metric):
2.1 mm
4.6 mm
Unit Size:
Pack of 3
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Diameter (Metric) unitSize
17125002 150 mm 2.1 mm Pack of 3
17155002 100 mm 4.6 mm Pack of 3
17165002 150 mm 4.6 mm Pack of 3
17115002 100 mm 2.1 mm Pack of 3
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 17125002 Supplier Waters Corporation Supplier No. 186008957

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

BioResolve RP mAb Columns contain superficially-porous-silica particles having an average pore diameter of 450 Å.

  • The particles are functionalized with a proprietary polyphenyl bonding process that was specifically designed for LC or LC-MS analyses of intact IgG monoclonal antibodies and their fragments.
  • This Method Validation Kit Part Number contains (3) columns from (3) different manufactured batches.

Specifications

Diameter (Metric) 2.1 mm
Length (Metric) 150 mm
Particle Shape Spherical
Particle Size 2.7 μm
Pore Size 450 Å
Quantity 3/Pk.
Type Method Validation Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.