missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Waters Corporation BioResolve-RP-mAb Column Method Validation Kit
Description
- The particles are functionalized with a proprietary polyphenyl bonding process that was specifically designed for LC or LC-MS analyses of intact IgG monoclonal antibodies and their fragments.
- This Method Validation Kit Part Number contains (3) columns from (3) different manufactured batches.
Specifications
Specifications
| Diameter (Metric) | 2.1 mm |
| Length (Metric) | 150 mm |
| Particle Shape | Spherical |
| Particle Size | 2.7 μm |
| Pore Size | 450 Å |
| Quantity | 3/Pk. |
| Type | Method Validation Kit |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?