Learn More
Neogen Clean-Trace™ Biomass Detection Kit ULW10
Product Code. 17218520
Encompass
Click to view available options
Quantity:
100/Cs.
Unit Size:
1 set
Product Code. 17218520
Brand: Neogen Clean-Trace™ 7000006881
This item requires a price update prior to being ordered.
Request a Price for this item here.
Water test kit using ATP bioluminescence to detect overall biological load in a sample.
  • Assesses the amount of biomass in process waters, waste waters, water tanks and cooling towers by determining the total ATP present.
  • Helps determine the biological status of process equipment and the effects of biocide treatments.
  • Useful for testing aqueous samples requiring filtration and/or extraction such as those that are viscous or that contain particulates or fibres.
Specifications
No. per Case 100 /Cs.
Quantity 100/Cs.
Type Test