Learn More
Thermo Scientific™ BIOLiner™ Rotor Bucket Adapters, Flat-bottom Bottle with Round Adapters
Combine Thermo Scientific™ BIOLiner™ Rotor Bucket Adapters with the BIOLiner Rotor Bucket for a complete system.
Brand:  Thermo Scientific™ 75003692
Product Code. 11854491
Description
Description
Combine Thermo Scientific™ BIOLiner™ Rotor Bucket Adapters with the BIOLiner Rotor Bucket for a complete system.
Ordering Information:
Individual adapters and bottle/adapter packages are offered.
  • Capacity range: 3L to 980mL
  • Maximum tube dimensions (O.D. x L): 98 × 124mm
Includes:
75-003-692 includes 750mL flat-bottom bottle with all round adapters for the TX-750 round bucket and four 500mL Corning™ conical bottle adapters.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Rotor Bucket Adapter
750 mL
750mL Flat-bottom Bottle w/Round Adapters for TX-750 Round Buckets
4