missing translation for 'onlineSavingsMsg'
Learn More

Alkali Scientific BioGuard™ Multi-layer Disposable Lab Coats

Product Code. 31297063
Encompass_Preferred
Change view
Click to view available options
Size:
Small
Medium
Large
3X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size
31297063 Small
31297062 Medium
31297061 Large
31297060 3X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 31297063 Supplier Alkali Scientific Supplier No. A8600S2X
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

These lab coats are made from breathable SMS fabric

  • Application: Laboratory and clinical settings where protection against fluids and particulate contamination is non-negotiable.
  • Advanced Protection: Crafted from premium SMS material, these lab coats offer an exceptional barrier against fluids and contaminants, keeping you safe in demanding work environments.
  • Anti-Static: Our premium lab coats are made from a cling-free material, unlike other lab coat materials.
  • Flame Retardant: Lab coats can be used in cleanrooms and other hazardous environments.
  • Breathable Design: The breathability of the SMS material ensures that you stay comfortable throughout your workday, preventing overheating or discomfort.
  • Safety First: Our lab coats are latex-free, making them safe for those with latex sensitivities or allergies.
  • Convenient Wear: Featuring a knit collar, cuffs, and a snap-button front, these lab coats are easy to put on and take off.
  • Practical Pockets: Three reinforced pockets provide convenient storage for essential tools and equipment.
  • Wide Size Range: Available in a range of sizes to accommodate a broad spectrum of users.
  • Bulk Quantity: Each order includes a case of 50 lab coats, ensuring you have an ample supply for your team.
  • Hanging Loop: The lab coats feature a practical hanging loop for easy storage and access.

Specifications

Size Small
Color Blue
Disposable Single-use
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.