Learn More
Cytiva (formerly Pall Lab) Biodyne™ C Membrane
Biodyne C membrane is negatively charged with pore surfaces populated by a high density of carboxyl groups.
Brand:  Cytiva (formerly Pall Lab) 60314
Additional Details : Weight : 0.00100kg
 View more versions of this product
Product Code. 17124511
  • £372.00 / Pack of 10
Estimated Shipment: 09-10-2024
to see stock.
Add to basket
Description
Description
  • The membrane can be derivatized by coupling reactions with these groups
  • It is ideal for reverse dot blots and affinity purification
Specifications
Specifications
Sheets
Negatively-charged Nylon 6.6
20 x 20 cm
Non-sterile
0.45 μm
11 to 13 μm