Learn More
Biodesign™ Biodesign™ Acrylic Levelling Platform
Description
10mm thick acrylic top with three individually adjustable chrome plated metal feet and a large easy to read bulls-eye level on the top.
Ensures a level surface when pouring and running polyacrylamide and agarose gels.
Ensures a level surface when pouring and running polyacrylamide and agarose gels.
Specifications
Specifications
| Type | Leveling Platform |