missing translation for 'onlineSavingsMsg'
Learn More

TerraCycle™ Biodegradable Plastic Zero Waste Box™

Product Code. 32016153
Change view
Click to view available options
Size:
Small
Medium
Large
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size
32016153 Large
32016152 Medium
32016151 Small
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 32016153 Supplier TerraCycle™ Supplier No. CS161LUK
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Support your sustainability goals and reduce landfill and incinerated waste with this Easy-to-use solution for industrial composting

  • Use this Zero Waste Box for: Biodegradable plastic Biodegradable 3D printing filaments, materials and scraps compostable plastic, PLA, PHA PLA products and packaging
  • Do not send in: Other types plastics (e.g. ABS, TPU) or products made from a mix of biodegradable plastic and other materials, food content, liquid waste, garden waste, organic matter, paper, wood and hazardous waste.
  • Each box is supplied with a Pre-affixed return shipping label.
    • Order: Select a Zero Waste Box™ and place an order.
    • Collect: Collect the accepted items to fill the box.
    • Ship: Once full, use the Pre-paid shipping label (already fastened to the box) to arrange for your waste to be collected for recycling.
    • Reorder: Order your next Zero Waste Box™ to continue to recycle.
  • The waste collected is processed at a compost facility instead of being sent to landfill or incineration.
  • Cardboard boxes are made from 100% recycled material.
  • Zero Waste Box is an All-inclusive Eco-friendly waste disposal service which includes storage, shipping and processing of the materials

Specifications

Capacity (Metric) 140 L
Certifications/Compliance FSC
Capacity (English) 37 gal.
Color Brown, Green
Dimensions (L x W x H) 38 x 38 x 97 cm
Disposable Single-use
Latex Free Yes
Size Large
Sterility Non-sterile
Autoclavable Not Autoclavable
Includes Return Shipping Label
For Use With (Application) Recycling Waste
Material Cardboard, Plastic
Product Type Recycling Box
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.