missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Falcon™ BioCoat™ 384-Well, Collagen Type I-Treated, Flat-Bottom Microplate



Click to view available options
Color:
Clear
Unit Size:
Pack of 5
Description
Ideal for applications involving adhesion, growth and differentiation of neuronal and transfected cell lines
- Source: rat tail
- TC-treated
- Clear, round well
- Flat bottom
- Sterile
- For research use onlyClosure: Yes For Use With (Application): Promotion of cell attachment and spreading, rapid expansion of cell populations, serum-free or reduced serum culture, cell adhesion assay, improving survival of primary cells in culture Sterile: No Surface Coating: ECM/Attachment Factors
Specifications
Specifications
| Certifications/Compliance | QC Tested for ability to promote attachment and spreading of HT-1080 human fibrosarcoma cells. Tested and found negative for bacteria and fungi. Collegen I purity >90% by SDS-PAGA. For research use only |
| Format | Standard |
| Lid | With Lid |
| Content And Storage | 4°C to 30°C under dry conditions |
| Array | 16 x 24 |
| Color | Clear |
| No. of Wells | 384 |
| Sterility | Sterile |
| Surface Treatment | Collagen Type I |
| Well Shape | Flat Bottom (F-well) |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction