missing translation for 'onlineSavingsMsg'
Learn More

Corning™ BioCoat™ 96-Well, Poly-D Lysine-Treated, Flat-Bottom Microplate
GREENER_CHOICEBIOPHARMA

Product Code. p-9045045
Click to view available options
Packaging:
5/Pk.
50/Cs
Unit Size:
Pack of 5
Pack of 50
This item is not returnable. View return policy

Product Code. 10379320

Brand: Corning™ 354651

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Color: White, Clear Bottom; Sterility: Sterile; Lid: With Lid

Poly-D-Lysine (PDL) and Poly-L-Lysine (PLL) are synthetic compounds that enhance cell adhesion and protein absorption by altering surface charges on the culture substrate. In addition to promoting cell adhesion, poly-lysine surface treatments support neurite outgrowth and improve the survival of many central nervous system (CNS) primary cells in culture. As PDL and PLL are synthetic molecules, they do not stimulate biological activity in the cells cultured on them, and they do not introduce impurities carried by natural polymers.

Applications include:

  • Attachment and spreading of a variety of cell types
  • Cell differentiation and neurite outgrowth
  • Attachment of fastidious transfected cell lines
  • Support survival of primary neurons in culture
  • Serum-free or reduced-serum culture

Source

  • PDL, synthetic (MW 75-150 kD)
  • PLL, synthetic (MW 30-70 kD)

Quality Control

  • Tested for ability to promote firm attachment of RCG cells
  • Tested and found negative for bacteria and fungi

Storage

  • 4°C to 30°C under dry conditions.
  • TRUSTED_SUSTAINABILITY

    Specifications

    Certifications/Compliance ANSI, SBS
    Format Standard
    Lid With Lid
    Content And Storage 4°C to 30°C
    Array 8 x 12
    Packaging 5/Pk.
    Color White, Clear Bottom
    Product Type Standard
    Material Polystyrene
    No. of Wells 96
    Sterility Sterile
    Surface Treatment Poly-D-Lysine
    Well Shape Round, Flat Bottom (F-well)
    For Use With (Application) Attachment and spreading of a variety of cell types, cell differentiation and neurite outgrowth, attachment of fastidious transfected cell lines, support survival of primary neurons in culture, serum-free or reduced serum culture
    No. per Case 5/Cs.
    No. per Pack 5/Pk.
    Quantity 5/Cs.
    Source Poly-D-Lysine
    Well Color Clear
    Show More Show Less
    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.