missing translation for 'onlineSavingsMsg'
Learn More

Corning™ BioCoat™ 96-Well, Poly-D Lysine-Treated, Flat-Bottom Microplate

Product Code. 10140403
Change view
Click to view available options
Packaging:
5/Cs
50/Cs
Unit Size:
Pack of 5
Pack of 50
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Packaging Unit Size
10140403 50/Cs Pack of 50
10043830 5/Cs Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10140403 Supplier Corning™ Supplier No. 356640
  • ONLINE EXCLUSIVE PRICE
    £960.00
    £892.63 / Pack of 50
    Save £67.37
    7% Off
Estimated Shipment: 04-02-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Color: Black, Clear Bottom; Sterility: Sterile; Lid: With Lid

Poly-D-Lysine (PDL) and Poly-L-Lysine (PLL) are synthetic compounds that enhance cell adhesion and protein absorption by altering surface charges on the culture substrate. In addition to promoting cell adhesion, poly-lysine surface treatments support neurite outgrowth and improve the survival of many central nervous system (CNS) primary cells in culture. As PDL and PLL are synthetic molecules, they do not stimulate biological activity in the cells cultured on them, and they do not introduce impurities carried by natural polymers.

Applications include:

  • Attachment and spreading of a variety of cell types
  • Cell differentiation and neurite outgrowth
  • Attachment of fastidious transfected cell lines
  • Support survival of primary neurons in culture
  • Serum-free or reduced-serum culture

Source

  • PDL, synthetic (MW 75-150 kD)
  • PLL, synthetic (MW 30-70 kD)

Quality Control

  • Tested for ability to promote firm attachment of RCG cells
  • Tested and found negative for bacteria and fungi

Storage

  • 4°C to 30°C under dry conditions.
  • TRUSTED_SUSTAINABILITY

    Specifications

    Certifications/Compliance ANSI, SBS
    Format Standard
    Lid With Lid
    Content And Storage 4°C to 30°C
    Array 8 x 12
    Packaging 50/Cs
    Color Black, Clear Bottom
    Material Polystyrene
    No. of Wells 96
    Sterility Sterile
    Surface Treatment Poly-D-Lysine
    Well Shape Round, Flat Bottom (F-well)
    For Use With (Application) Attachment and spreading of a variety of cell types, cell differentiation and neurite outgrowth, attachment of fastidious transfected cell lines, support survival of primary neurons in culture, serum-free or reduced serum culture
    No. per Case 50/Cs.
    No. per Pack 5/Pk.
    Quantity 50/Cs.
    Source Poly-D-Lysine
    Well Color Clear
    Show More Show Less
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.