missing translation for 'onlineSavingsMsg'
Learn More

Corning™ BioCoat™ 96-Well, Collagen Type I-Treated, Flat-Bottom Microplate
GREENER_CHOICEBIOPHARMA

Catalog Number p-9045039
Click to view available options
Packaging:
5/Cs
5/Pk., 50/Cs.
80/Cs

Color: Clear; Sterility: Sterile; Lid: With Lid

  • Source: Rat tail
  • TC-treated
  • Clear, round well
  • Flat bottom
  • Sterile

For research use only.

TRUSTED_SUSTAINABILITY
Certifications/Compliance ANSI, SBS
Format Standard
Lid With Lid
Content And Storage 4°C to 30°C
Culture Area 0.32 cm2
Array 8 x 12
Packaging 5/Cs
Color Clear
Material Polystyrene
No. of Wells 96
Sterility Sterile
Surface Treatment Collagen Type I
Well Shape Round, Flat-bottom
For Use With (Application) Promotion of cell attachment and spreading, rapid expansion of cell populations, serum-free or reduced serum culture, cell adhesion assay, improving survival of primary cells in culture
No. per Case 5/Cs.
No. per Pack 5/Pk.
Quantity 5/Cs.
Source Rat Tail
Well Color Clear
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'