Learn More
Ansell BIOCLEAN™ Permaflow Irradiated Cleanroom Ballpoint Pen S-BPBP-1
Product Code. p-200080449
Click to view available options
Color:
Black
Blue
Unit Size:
Pack of 300
Product Code. 30040334
Brand: Ansell SBPBP1BK
Only null left
Add to basket
Add to basket
Ballpoint pens for alcohol resistant labelling are processed to ensure ISO Class 4 and EU GMP Grade A compatibility, fade and water resistant, quick drying, and non-toxic.
  • BioClean Permaflow cleanroom pens have fade and water resistant fast drying permanent ink, and feature an alcohol resistant label for easy cleaning within a critical environment, they are ISO Class 4 and EU GMP Grade A compatible.
  • With a robust, lightweight, tinted polypropylene round barrel and 1 mm point size for quality writing precision. Ink complies with documentary use standard ISO 12757-2
TRUSTED_SUSTAINABILITY
Specifications
Tip Style Rollerball
Product Type Ballpoint Pen
Color Black