missing translation for 'onlineSavingsMsg'
Learn More

Ansell BioClean™ S-BGHF Sterile Half-Finger Nylon Glove Liners

Product Code. 15248308
Change view
Click to view available options
One Size
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
15248308 One Size
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15248308 Supplier Ansell Supplier No. SBGHF

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Glove liners are designed specifically to provide a comfortable barrier between the skin and the outer glove. Suitable for use in Class 10/ISO 4 Cleanrooms.

  • Half-finger style for maximum tactility
  • Provides a soft barrier between the skin and glove
  • EU GMP Grade A
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance (Europe) EN 420:2003 + A1:2009
Cleanroom Class Class 10 (ISO 4)
Coating Material Uncoated
Color White
Hand Specific
Latex Free Yes
Material Nylon
Size Universal
Sterility Sterile
Finger Style Half
For Use With (Application) Life Sciences
Glove Cuff Style Beaded
Series BGHF
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.