Learn More
Ansell BioClean™ Durableu Disposable Overboots BDBL
Description
- Resists tears, rips and abrasions
- Low levels of particle shedding
- Resistant to a wide range of liquid chemicals
- High tensile strength
- Slip-resistant
- 102gsm non-woven, spunbonded Polypropylene (38gsm)/Cast Polyethylene (64gsm) laminate
Specifications
Specifications
| Size | Universal |
| Latex Free | No |
| Color | Blue |
| Disposable | Single-use |
| Sterility | Non-sterile |
| Certifications/Compliance | Manufactured in a facility holding ISO 9001:2015 |
| Packaging Quantity | 50 pieces per sealed inner PE bag; one inner bag per sealed outer PE bag; eight outer bags per lined carton (400 pieces) |
| Resistance | Chemical-resistant |
| Product Type | Controlled Environments Boot Covers |
Five (5) years from date of manufacture. Store in a dry, cool place (≤40°C) away from direct sunlight and fluorescent light.