missing translation for 'onlineSavingsMsg'
Learn More

Ansell BioClean™ Sterile Dual Overshoes
BIOPHARMA

Product Code. 17801055
Change view
Click to view available options
Closure Color:
White
Unit Size:
150 pairs
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Closure Color Unit Size
17801055 White 150 pairs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17801055 Supplier Ansell Supplier No. SBDOS
Support a Safer, More Efficient Laboratory
Use promo code 28026
View all promotions
Estimated Shipment: 02-06-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Disposable overshoes are practical and durable, featuring a heavy-duty textured cast polyethylene slip-resistant sole for a secure footing.

  • Non-woven spunbonded polypropylene
  • Heavy duty slip-resistant sole
  • Latex-free elastic
  • Manufactured in a facility holding ISO 9001:2008 certification.
  • Processed in an NEBB certified ISO Class 4 environment
TRUSTED_SUSTAINABILITY

Specifications

Material Polypropylene
Latex Free Yes
Color White
Disposable Single-use
Sterility Sterile
Closure Type Elastic Ankle
Sole Type Anti-slip
Closure Color White
For Use With (Application) Life Sciences
Packaging Quantity 150 Pairs/Cs.
Length (English) 16 in.
Length (Metric) 400 mm
Height (Metric) 152 mm
Product Type Controlled Environments Shoe Covers
Cleanroom Class Class 10 (ISO 4)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.