Learn More
Ansell BioClean-D™ Sterile Hood
BIOPHARMA
Product Code. 16673622
Change view
Click to view available options
Material:
Polypropylene
Unit Size:
Pack of 120
Product Code. Material Unit Size
16673622 Polypropylene Pack of 120
1 options
N/A
Product Code. 16673622 Supplier Ansell Supplier No. SBDHDL
Support a Safer, More Efficient Laboratory
Use promo code 28026
View all promotions
Estimated Shipment: 23-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
N/A
Long length cleanroom hood features an extra-long yoke for maximum coverage.
  • Extended length: More complete protective coverage
  • Low-linting lightweight material: Fewer contamination risks
  • ESD properties: Reduced risk of electrostatic interference or damage
TRUSTED_SUSTAINABILITY
Specifications
Product Type Sterile Hood
Size Universal
Material Polypropylene
Color White
Closure Type Pull-over
Disposable Single-use
Latex Free No
Model S-BDHD
Seam Type Bound
Size (English) Universal
Size (Metric) Universal
Sterility Sterile
Construction Low-linting, Lightweight Material
For Use With (Application) Laboratory, Crime Scene Investigation (CSI), Forensics, Cleanroom, Medical Device Manufacturing, Biotechnology, Maintaining Equipment and Instruments, Pharmaceutical
Resistance Electrostatic Discharge
Cleanroom Class Class 10 (ISO 4)
Show More Show Less
N/A