Learn More
Novacyt - Microgen Bioproducts Biochemical Identification System: Enterobacteriacae
Product Code. 15987112 Shop All Novacyt Group Products
Click to view available options
Quantity:
24 Tests
60 Tests
Unit Size:
24 tests
60 tests
Product Code. 15987112
Brand: Novacyt - Microgen Bioproducts MID65CE
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Biochemical identification system for Enterobacteriaceae from foods and other environmental samples.
  • Twelve-substrate biochemical identification system.
  • From a range of different technologies for the specific detection of Enterobacteriaceae from foods and other environmental samples.
  • From a range of simple, reliable, bacterial identification systems for food, environmental, and industrial laboratories.
  • These tests will arm professionals with the necessary information to maximize the effectiveness of appropriate patient management.
  • The range is expanding quickly and will encompass the widest range of microbiological testing solutions.
Specifications
For Use With (Application) Identification of Enterobacteriaceae
Quantity 24 Tests