missing translation for 'onlineSavingsMsg'
Learn More

Simport™ Scientific BioBlock™ Deep Well Plates

Product Code. 11670771
Change view
Click to view available options
Color:
Pink
Unit Size:
Pack of 24
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color unitSize
11670771 Pink Pack of 24
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Product Code. 11670771 Supplier Simport™ Scientific Supplier No. T1105P
Only null left
Add to basket
Edge
Add to basket
This item is not returnable. View return policy
You may only order up to the available stock. Where an alternative item has been identified it will be shown below

These rugged plates are compatible with all leading robotic sample processors, automated liquid handling systems and 8- and 12- channel pipettors.

  • Conform to the SBS standard footprint and are identical in size to 96-well microtitre plates
  • Will withstand temperatures up to 121 °C
  • Fully compatible with deep freezing work, down to -196 °C
  • Alphanumeric grid for sample identification
  • Stackable
  • Withstand centrifugation up to 6000xg by using microtitre plate rotors
  • DMSO resistant
  • Designed for high-throughput screening, well suited for combinatorial chemistry
  • Ideal for sample storage and automated plate pipetting
  • Easy to empty completely and ideal for culturing bacterial cells as well as general procedures requiring a mother plate, DNA sequencing, ELISA, etc...

Specifications

Array 8 x 12
Color Pink
Product Type 96-well Deep well plate
For Use With (Equipment) Robotic Sample Processors, Automated Liquid Handling Systems and 8- and 12- Channel Pipettors
Material Polypropylene
No. of Wells 96
Sterility Non-sterile
Total Volume (Metric) Well 1.2 mL
For Use With (Application) Deep Freezing Work; High-Throughput Screening; Combinatorial Chemistry; Sample Storage; Automated Plate Pipetting; General Procedures Requiring a Mother Plate; DNA Sequencing; ELISA; Cultering Bacterial Cells; Etc...
Max. RCF 6000 xg (by using microtitre plate rotors)
Temperature Range (Metric) -196°C to +121°C
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.