Learn More
Thermo Scientific™ BioBasic™ PREP SEC 60 Size Exclusion Chromatography HPLC Columns
Achieve outstanding size separation of water soluble compounds with BioBasic™ PREP SEC 60 Size Exclusion Chromatography HPLC Columns.
£635.22 - £1967.03
Specifications
Max. Pressure | 4300 psi (295 bar) |
---|---|
Particle Size | 5 μm |
pH | 2 to 8 |
Pore Size | 60 Å |
Temperature | 60°C |
Product Code | Brand | Length (Metric) | Column Format | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Column Format | Price | Quantity & Availability | ||||
16631674
|
Thermo Scientific™
73305-037821A |
30 mm | Preparative Guard Column |
£635.22
Each |
Estimated Shipment: 12-06-2024
Log in to see stock. |
||||
16641674
|
Thermo Scientific™
73305-157846A |
150 mm | Preparative Column, Standard |
£1315.90
Each |
Estimated Shipment: 13-06-2024
Log in to see stock. |
||||
16506830
|
Thermo Scientific™
73305-307846A |
300 mm | Preparative Column, Standard |
£1967.03
Each |
Estimated Shipment: 20-06-2024
Log in to see stock. |
||||
Description
Provide outstanding size separation of water soluble compounds with Thermo Scientific™ BioBasic™ SEC 60 Size Exclusion Chromatography preparative HPLC Columns. BioBasic SEC columns provide high efficiency separations for a wide range of molecular weights from 100 to 60,000 daltons, allowing for the analysis of proteins, peptides and other biomolecules.
BioBasic™ PREP SEC 60 HPLC Column Benefits:
- Covers separation of analytes over a wide molecular weight range
- Long column life and high column efficiencies
- Simple mechanism of interaction based on molecular size and shape
- Excellent for sample cleanup
- Straightforward method development, simple mobile phases
- 60Å pore size
Ordering Information:
For information on bulk quantities, please inquire.
Specifications
4300 psi (295 bar) | |
2 to 8 | |
60°C | |
Spherical, Fully Porous, Ultrapure Silica | |
Size Exclusion | |
Size Exclusion |
5 μm | |
60 Å | |
Diol (OH) | |
>9% | |
No | |
Preparative Chromatography |