missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ BioBasic™ PREP SEC 1000 Size Exclusion Chromatography HPLC Columns

Product Code. 16691674 Shop All Thermo Scientific Products
Change view
Click to view available options
Length (Metric):
30 mm
150 mm
300 mm
Unit Size:
Each
Product Code. Length (Metric) unitSize
16691674 30 mm Each
16601684 150 mm Each
16546130 300 mm Each
3 options
This item is not returnable. View return policy

Product Code. 16691674

Brand: Thermo Scientific™ 73605037821A

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Achieve outstanding size separation of water soluble compounds with BioBasic™ PREP SEC 1000 Size Exclusion Chromatography HPLC Columns.

Provide outstanding size separation of water soluble compounds with Thermo Scientific™ BioBasic™ SEC 1000 Size Exclusion Chromatography preparative HPLC Columns. BioBasic SEC columns provide high efficiency separations for a wide range of molecular weights from 70,000 to 10,000,000 daltons, allowing for the analysis of proteins, peptides and other biomolecules.

BioBasic™ PREP SEC 120 HPLC Column Benefits:

  • Covers separation of analytes over a wide molecular weight range
  • Long column life and high column efficiencies
  • Simple mechanism of interaction based on molecular size and shape
  • Excellent for sample cleanup
  • Straightforward method development, simple mobile phases
  • 120Å pore size

Ordering Information:

For information on bulk quantities, please inquire.

TRUSTED_SUSTAINABILITY

Specifications

Column Format Preparative Guard Column
Diameter (Metric) 7.8 mm
For Use With Preparative Size Exclusion
Length (Metric) 30 mm
Max. Pressure 4300 psi (295 bar)
Particle Size 5 μm
pH 2 to 8
Surface Area 25 m²/g
Temperature 60°C
USP Type L59
Stationary Phase Diol (OH)
Packing Material Spherical, Fully Porous, Ultrapure Silica
Carbon Load >0.6%
Column Type Size Exclusion
Endcapped No
Phase Size Exclusion
For Use With (Application) Preparative Chromatography
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.