missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ BioBasic™ AX HPLC Columns

Product Code. 11598330 Shop All Thermo Scientific Products
Change view
Click to view available options
Length (Metric):
10 mm
50 mm
100 mm
150 mm
250 mm
Diameter (Metric):
1 mm
2.1 mm
3 mm
4 mm
4.6 mm
Unit Size:
Each
Pack of 4
16 product options available for selection
Product selection table with 16 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Diameter (Metric) unitSize
11598330 10 mm 4 mm Pack of 4
10552465 50 mm 1 mm Each
10682284 50 mm 2.1 mm Each
10446644 50 mm 3 mm Each
10304183 50 mm 4.6 mm Each
10519945 100 mm 1 mm Each
10314183 100 mm 2.1 mm Each
10513415 100 mm 4.6 mm Each
10170703 150 mm 1 mm Each
10671324 150 mm 2.1 mm Each
10221005 150 mm 3 mm Each
10630754 150 mm 4.6 mm Each
10253603 250 mm 4.6 mm Each
11578330 10 mm 1 mm Pack of 4
11588330 10 mm 2.1 mm Pack of 4
10659675 10 mm 3 mm Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 16 options available.
16 options
This item is not returnable. View return policy
Product Code. 11598330 Supplier Thermo Scientific™ Supplier No. 73105014001

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Use Thermo Scientific™ BioBasic™ AX LC Columns for the separation of proteins, peptides, other anionic species and highly polar molecules.

Use Thermo Scientific™ BioBasic™ AX LC Columns for the separation of proteins, peptides, other anionic species and highly polar molecules.

  • 5μm particle size, 300Å pore size
  • Proprietary, protein-friendly, ion exchange ligands of polyethyleneimine (PEI) bound to pure deactivated silica
  • Low hydrophobicity
  • Capacity of 0.22mEq/g; 3% carbon load
  • Use for normal phase and hydrophilic interaction chromatography (HILIC)
  • Stable under demanding pH conditions (2 to 8 pH range)
TRUSTED_SUSTAINABILITY

Specifications

Column Format Guard Cartridge, 4 Pk.
Diameter (Metric) 4 mm
Length (Metric) 10 mm
Max. Pressure 5800 psi (400 bar)
Particle Size 5 μm
pH 2 to 8
Pore Size 300 Å
Surface Area 100 m²/g
Temperature 60°C
Stationary Phase Polyethyleneimine
Product Line BioBasic
Packing Material Deactivated Silica
Carbon Load 3%
Column Type Mixed Mode
Endcapped No
Phase Normal Phase, HILIC, Ion Exchange
Type Guard Cartridge, 4 Pk.
Technique Anion Exchange
Show More Show Less

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.