missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ BioBasic™ SCX HPLC Columns

Product Code. 10344423 Shop All Thermo Scientific Products
Change view
Click to view available options
Length (Metric):
10 mm
50 mm
100 mm
150 mm
250 mm
Diameter (Metric):
1 mm
2.1 mm
3 mm
4 mm
4.6 mm
Unit Size:
Each
Pack of 4
18 product options available for selection
Product selection table with 18 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Diameter (Metric) unitSize
10344423 50 mm 2.1 mm Each
10150943 50 mm 3 mm Each
10323105 100 mm 1 mm Each
10562455 100 mm 2.1 mm Each
10271275 100 mm 3 mm Each
10354133 100 mm 4.6 mm Each
10538205 150 mm 1 mm Each
10602294 150 mm 2.1 mm Each
10699993 150 mm 3 mm Each
10181123 150 mm 4.6 mm Each
10496654 250 mm 1 mm Each
10612294 250 mm 2.1 mm Each
10572265 250 mm 4.6 mm Each
10725564 50 mm 4.6 mm Each
10331115 10 mm 1 mm Pack of 4
11538340 10 mm 4 mm Pack of 4
10545625 10 mm 3 mm Pack of 4
11528340 10 mm 2.1 mm Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 18 options available.
18 options
This item is not returnable. View return policy
Product Code. 10344423 Supplier Thermo Scientific™ Supplier No. 73205052130
Estimated Shipment: 31-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

BioBasic SCX LC Columns are optimized for the separation of proteins, peptides and other cationic species.

Use Thermo Scientific™ BioBasic™ SCX LC Columns for the optimized separation of proteins, peptides and other cationic species. These high-performance columns are ideal for the analysis of proteins, peptides and other biomolecules with higher efficiency than polymeric ion exchange.

  • Strong cation exchange phase based on sulfonic acid chemistry
  • Separation and retention of basic and other cationic species
  • 300Å pore size for enhanced protein and peptide separations
  • Outstanding stability under demanding pH conditions

Ordering Information:

Other column dimensions are available. Please call Customer Service for more information.

TRUSTED_SUSTAINABILITY

Specifications

Column Format Analytical Column
Diameter (Metric) 2.1 mm
Length (Metric) 50 mm
Max. Pressure 5800 psi (400 bar)
Particle Size 5 μm
pH 2 to 8
Pore Size 300 Å
Surface Area 100 m²/g
Temperature 60°C
USP Type L52
Stationary Phase Sulfonic Acid
Product Line BioBasic
Packing Material Spherical, Fully Porous, Ultrapure Silica
Carbon Load 3%
Column Type Ion Exchange
Endcapped No
Phase Ion Exchange
Product Type Analytical Column
Show More Show Less

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.