missing translation for 'onlineSavingsMsg'
Learn More

BioAcryl-P (30%, 29:1)

Product Code. 15475549
Change view
Click to view available options
Quantity:
500 mL
1 L
Unit Size:
1 L
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15475549 1 L 1 L
15465549 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15475549 Supplier Thermo Scientific Alfa Aesar Supplier No. J63279.K2
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

BioAcryl-P (Acrylamide/Bisacrylamide 29:1, CAS # 25034-58-6) is a powder-based formula used to create the gel matrix necessary for electrophoresis experiments.

Acrylamide is used for sequencing gels and separation of proteins, used in protein and nucleic acid electrophoresis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• BioAcryl-P is a 29:1 ratio of Acrylamide/Bisacrylamide
• Acrylamide is a crystalline amide that forms polymer structures at room temperature
• Acrylamide acts as the monomer for the gel structure, and bisacrylamide as the crosslinker for gel matrix formation

Application

• Suitable for gel electrophoresis-mediated separation of proteins and DNA for identification or sequencing

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Acrylamide/Bisacrylamide 29:1
Physical Form Liquid
Concentration 30% soln
Color Colorless
Quantity 1 L
MDL Number MFCD00080848
UN Number UN3426
Solubility Information Difficult to mix.
compliance-icons compliance-icons
Product Identifier
  • BioAcryl-P (30%, 29:1)
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Carcinogenicity Category 1B
  • Serious eye damage/eye irritation Category 2
  • Germ cell mutagenicity Category 1B
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H340-May cause genetic defects.
  • H350-May cause cancer.
  • H361f-Suspected of damaging fertility.
  • H372-Causes damage to organs.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: Acrylamide, Methylene diacrylamide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.