missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies BIO
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
Description
- BIO is an indirubin compound that is cell-permeable and is a selective and potent reversible inhibitor of GSK3α and GSK3β (IC50 = 5 nM) and functions as a WNT activator
- Enhances reprogramming of mouse fibroblasts, neural stem cells, and thymocytes to induced pluripotent stem (iPS) cells (Lluis et al.)
- Maintains self-renewal of mouse embryonic stem (ES) cells in the absence of LIF (Sato et al.)
- Maintains self-renewal of human ES cells in unconditioned medium through a SMAD 2/3 dependent mechanism (James et al.), Sato et al.)
- Induces proliferation of neonatal and adult rat cardiomyocytes (Tseng et al.)
- Promotes differentiation of cardiomyocytes from human ES and iPS cells (Lian et al.)
Specifications
Specifications
| CAS | 667463-62-9 |
| Synonym | GSK 3 IX; MLS 2052; 6-Bromoindirubin-3'-oxime |
| Quantity | 1 mg |
| Molecular Formula | C16H10BrN3O2 |
| Purity | ≥98% |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction