Learn More
Olympus™ Binocular Microscope, Basic Stand, LED Illumination, SZ51
Product Code. 15566535
Change view
Click to view available options
Head Style:
Binocular
Unit Size:
1 unit
Product Code. Head Style Unit Size
15566535 Binocular 1 unit
1 options
Product Code. 15566535 Supplier Olympus™ Supplier No. K23008480
  • ONLINE EXCLUSIVE PRICE
    £4282.90
    £1928.00 / Pack of 1
    Save £2354.90
    54% Off
Estimated Shipment: 20-01-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Deliver images with superb depth, clarity, and detail. Olympus™ Binocular Microscope, Basic Stand, LED Illumination, SZ51 uses dependable, high-performance optics to produce consistent and accurate results and a V-shape optical path to ensure a slim body ideal for equipment integration or for stand alone use.
  • LED transmitted light with mirror contrast adjustment.
  • Binocular head, 45º inclination.
  • Interpupillary adjustment 52-76mm.
  • Greenough zoom optical system.
  • Widefield eyepieces WF10X FN22mm.
  • Zoom ratio 5:1.
  • 110mm working distance.
  • Stand with focusing & ESD capability.
  • Black and white stage plate
Specifications
Product Type Microscope
Eyepiece WF 10X FN22 mm
Head Style Binocular
Lighting Type LED
Magnification Power 0.8X to 4X