missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Binding Buffer NT

Product Code. 15829107
Change view
Click to view available options
Quantity:
1000 mL
Unit Size:
1000mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15829107 1000 mL 1000mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15829107 Supplier Macherey-Nagel™ Supplier No. 740614.1
  • ONLINE EXCLUSIVE PRICE
    £152.00
    £139.57 / 1000mL
    Save £12.43
    8% Off
 Restricted Item
Estimated Shipment: 21-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Binding Buffer NT to be used with NucleoSpin 8 / 96 PCR Clean‐up kits

Specifications

Chemical Name or Material Binding buffer NT
CAS 593-84-0
Color Colorless
For Use With (Application) Direct clean-up of PCR reaction mixtures
For Use With (Equipment) NucleoSpin™ PCR Clean-up kits
pH 4.5 to 5.5
Physical Form Liquid
Quantity 1000 mL
Formulation Guanidinium thiocyanate
Odor Acetic
compliance-icons
Product Identifier
  • Buffer NT
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H412-Harmful to aquatic life with long lasting effects.
Supplemental information
  • MIXTURE LIST-Contain: guanidinium thiocyanate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.