missing translation for 'onlineSavingsMsg'
Learn More

Bindarit, MedChemExpress

Product Code. 16444377
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16444377 50 mg 50mg
16438508 10 mg 10mg
16438346 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16444377 Supplier Medchem Express Supplier No. HYB0498

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Bindarit (AF2838) is a selective inhibitor of the monocyte chemotactic proteins MCP-1/CCL2, MCP-3/CCL7, and MCP-2/CCL8, and no effect on other CC and CXC chemokines such as MIP-1α/CCL3, MIP-1β/CCL4, MIP-3/CCL23. Bindarit also has anti-inflammatory activity.

Bindarit (AF2838) is a selective inhibitor of the monocyte chemotactic proteins MCP-1/CCL2, MCP-3/CCL7, and MCP-2/CCL8, and no effect on other CC and CXC chemokines such as MIP-1α/CCL3, MIP-1β/CCL4, MIP-3/CCL23. Bindarit also has anti-inflammatory activity.

Specifications

Chemical Name or Material Bindarit
Physical Form Solid
CAS 130641-38-2
Color White
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C19H20N2O3
For Use With (Application) COVID-19-immunoregulation
Synonym AF2838
Solubility Information DMSO : ≥ 46 mg/mL (141.81 mM)
SMILES CC(OCC1=NN(CC2=CC=CC=C2)C3=C1C=CC=C3)(C)C(O)=O
Molecular Weight (g/mol) 324.37
Formula Weight 324.37
Percent Purity 99.47%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.