missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BIN3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11215-1-AP-150UL
This item is not returnable.
View return policy
Description
Involved in cytokinesis and septation where it has a role in the localization of F-actin.Specifications
| BIN3 | |
| Polyclonal | |
| Unconjugated | |
| BIN3 | |
| BIN3, bridging integrator 3 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 361065, 55909, 57784 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q68FW8, Q9JI08, Q9NQY0 | |
| Bin3 | |
| BIN3 Fusion Protein Ag1713 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction