missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BIN3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35560-100ul
This item is not returnable.
View return policy
Description
BIN3 Polyclonal antibody specifically detects BIN3 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
| BIN3 | |
| Polyclonal | |
| Western Blot 1:200 - 1:2000, ELISA | |
| bridging integrator 3, MGC14978 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-253 of human BIN3 (NP_061158.1).,, Sequence:, MSWIPFKIGQPKKQIVPKTVERDFEREYGKLQQLEEQTRRLQKDMKKSTDADLAMSKSAVKISLDLLSNPLCEQDQDLLNMVTALDTAMKRMDAFNQEKVNQIQKTVIEPLKKFGSVFPSLNMAVKRREQALQDYRRLQAKVEKYEEKEKTGPVLAKLHQAREELRPVREDFEAKNRQLLEEMPRFYGSRLDYFQPSFESLIRAQVVYYSEMHKIFGDLSHQLDQPGHSDEQRERENEAKLSELRALSIVADD | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 55909 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction