missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BIN2 Antibody, mFluor Violet 450 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97015MFV450
This item is not returnable.
View return policy
Description
BIN2 Polyclonal antibody specifically detects BIN2 in Human samples. It is validated for Western Blot, ELISA
Specifications
| BIN2 | |
| Polyclonal | |
| Western Blot, ELISA | |
| BRAP1, BRAP-1, breast cancer associated protein BRAP1, Breast cancer-associated protein 1, bridging integrator 2, bridging integrator-2 | |
| Produced in rabbits immunized with purified, recombinant Human BIN2 (Uniprot#: Q9UBW5-1; Met1-Asn244). | |
| 0.1 mL | |
| Cell Cycle and Replication | |
| 51411 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, ELISA | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction