missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BIN1 Mouse anti-Human, Mouse, Clone:3B6F10, Abnova™
Mouse monoclonal antibody raised against recombinant human BIN1.
Brand: Abnova MAB17313.100ug
This item is not returnable.
View return policy
Description
This gene encodes several isoforms of a nucleocytoplasmic adaptor protein, one of which was initially identified as a MYC-interacting protein with features of a tumor suppressor. Isoforms that are expressed in the central nervous system may be involved in synaptic vesicle endocytosis and may interact with dynanim, synaptojanin, endophilin, and clathrin. Isoforms that are expressed in muscle and ubiquitously expressed isoforms localize to the cytoplasm and nucleus and activate a caspase-independent apoptotic process. Studies in mouse suggest that this gene plays an important role in cardiac muscle development. Alternate splicing of the gene results in ten transcript variants encoding different isoforms. Aberrant splice variants expressed in tumor cell lines have also been described. [provided by RefSeq]Specifications
| BIN1 | |
| Monoclonal | |
| Unconjugated | |
| ELISA (1:10000) Flow Cytometry (1:200-400) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:200-1000) Western Blot (1:500-1:2000) The optimal working dilution should be determined by the end user. |
|
| BIN1 | |
| Mouse | |
| 100 μg | |
| Human, Mouse | |
| Antibody | |
| IgG2b |
| ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| 3B6F10 | |
| bridging integrator 1 | |
| In PBS (0.05% sodium azide) | |
| AMPH2, AMPHL, DKFZp547F068, MGC10367, SH3P9 | |
| Recombinant protein corresponding to amino acids 189-398 of of human BIN1 from E. coli. | |
| 274 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction