missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BIN1 Antibody (1H1), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals H00000274-M01
This item is not returnable.
View return policy
Description
BIN1 Monoclonal antibody specifically detects BIN1 in Human samples. It is validated for Western Blot, ELISA
Specifications
| BIN1 | |
| Monoclonal | |
| Unconjugated | |
| In 1x PBS, pH 7.4 | |
| AMPH2, Amphiphysin II, Amphiphysin-like protein, AMPHLDKFZp547F068, box dependant MYC interacting protein 1, bridging integrator 1Box-dependent myc-interacting protein 1, MGC10367, myc box-dependent-interacting protein 1, SH3P9 | |
| BIN1 (NP_004296, 355 a.a. ~ 454 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. VVETFPATVNGTVEGGSGAGRLDLPPGFMFKVQAQHDYTATDTDELQLKAGDVVLVIPFQNPEEQDEGWLMGVKESDWNQHKELEKCRGVFPENFTERVP | |
| 0.1 mg | |
| Cell Cycle and Replication, Tumor Suppressors | |
| 274 | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
| IgG1 κ |
| Western Blot, ELISA | |
| 1H1 | |
| Western Blot 1:500, ELISA | |
| NP_004296 | |
| Mouse | |
| IgG purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction