missing translation for 'onlineSavingsMsg'
Learn More

Bimiralisib, MedChemExpress

Product Code. 16445418
Encompass_Preferred
Click to view available options
Quantity:
2 mg
5 mg
10 mg
100 mg
Unit Size:
100mg
10mg
2mg
5mg
This item is not returnable. View return policy

Product Code. 16445418

Brand: Medchem Express HY12868

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Bimiralisib (PQR309) is a potent, brain-penetrant, orally bioavailable, pan-class I PI3K/mTOR inhibitor with IC50s of 33 nM, 451 nM, 661 nM, 708 nM and 89 nM for PI3Kα, PI3Kδ, PI3Kβ, PI3Kγ and mTOR, respectively. Bimiralisib is an mTORC1 and mTORC2 inhibitor.

Bimiralisib (PQR309) is a potent, brain-penetrant, orally bioavailable, pan-class I PI3K/mTOR inhibitor with IC50s of 33 nM, 451 nM, 661 nM, 708 nM and 89 nM for PI3Kα, PI3Kδ, PI3Kβ, PI3Kγ and mTOR, respectively. Bimiralisib is an mTORC1 and mTORC2 inhibitor.

Specifications

Chemical Name or Material Bimiralisib
CAS 1225037-39-7
Color Gray
Purity Grade Notes Research
Molecular Formula C17H20F3N7O2
Quantity 10 mg
Synonym PQR309
Solubility Information DMSO : ≥ 50 mg/mL (121.54 mM)
SMILES NC1=NC=C(C2=NC(N3CCOCC3)=NC(N4CCOCC4)=N2)C(C(F)(F)F)=C1
Molecular Weight (g/mol) 411.38
Formula Weight 411.38
Percent Purity 98.74%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.