missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bimiralisib, MedChemExpress
missing translation for 'orderingAttributeHoverText'
Quantity:
2 mg
5 mg
10 mg
100 mg
missing translation for 'unitSize'
100mg
10mg
2mg
5mg
Description
Bimiralisib (PQR309) is a potent, brain-penetrant, orally bioavailable, pan-class I PI3K/mTOR inhibitor with IC50s of 33 nM, 451 nM, 661 nM, 708 nM and 89 nM for PI3Kα, PI3Kδ, PI3Kβ, PI3Kγ and mTOR, respectively. Bimiralisib is an mTORC1 and mTORC2 inhibitor.
Specifications
Specifications
| Chemical Name or Material | Bimiralisib |
| CAS | 1225037-39-7 |
| Color | Gray |
| Purity Grade Notes | Research |
| Molecular Formula | C17H20F3N7O2 |
| Quantity | 2 mg |
| Synonym | PQR309 |
| Solubility Information | DMSO : ≥ 50 mg/mL (121.54 mM) |
| SMILES | NC1=NC=C(C2=NC(N3CCOCC3)=NC(N4CCOCC4)=N2)C(C(F)(F)F)=C1 |
| Molecular Weight (g/mol) | 411.38 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction