missing translation for 'onlineSavingsMsg'
Learn More

Bimatoprost, TRC

Product Code. 17952475
Change view
Click to view available options
Quantity:
1 mg
250 mg
1 g
Unit Size:
1g
1mg
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17952475 250 mg 250mg
17914515 1 g 1g
17992575 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17952475 Supplier Toronto Research Chemicals Supplier No. TRCB386800250MG
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 155206-00-1
Molecular Formula C25 H37 N O4
Molecular Weight (g/mol) 415.57
Synonym (5Z)-7-[(1R,2R,3R,5S)-3,5-Dihydroxy-2-[(1E,3S)-3-hydroxy-5-phenyl-1-penten-1-yl]cyclopentyl]-N-ethyl-5-heptenamide, 5-Heptenamide, 7-[(1R,2R,3R,5S)-3,5-dihydroxy-2-[(1E,3S)-3-hydroxy-5-phenyl-1-pentenyl]cyclopentyl]-N-ethyl-, (5Z)- (9CI), 5-Heptenamide, 7-[3,5-dihydroxy-2-(3-hydroxy-5-phenyl-1-pentenyl)cyclopentyl]-N-ethyl-, [1R-[1α(Z),2β(1E,3S*),3α,5α]]-, 17-Phenyltrinor-PGF2α ethylamide, AGN 192024, Bimatoprost, Latisse, Lumigan, Prostamide
IUPAC Name (Z)-7-[(1R,2R,3R,5S)-3,5-dihydroxy-2-[(E,3S)-3-hydroxy-5-phenylpent-1-enyl]cyclopentyl]-N-ethylhept-5-enamide
SMILES CCNC(=O)CCC\C=C/C[C@H]1[C@@H](O)C[C@@H](O)[C@@H]1\C=C\[C@@H](O)CCc2ccccc2

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C25H37NO4/c1-2-26-25(30)13-9-4-3-8-12-21-22(24(29)18-23(21)28)17-16-20(27)15-14-19-10-6-5-7-11-19/h3,5-8,10-11,16-17,20-24,27-29H,2,4,9,12-15,18H2,1H3,(H,26,30)/b8-3-,17-16+/t20-,21+,22+,23-,24+/m0/s1
Formula Weight 415.2723
Percent Purity >95%
Chemical Name or Material Bimatoprost
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.