missing translation for 'onlineSavingsMsg'
Learn More

Bimatoprost, MedChemExpress

Product Code. 16478398
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16478398 10 mg 10mg
16470808 5 mg 5mg
16464287 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16478398 Supplier Medchem Express Supplier No. HYB0191
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Bimatoprost is a prostaglandin analog used topically (as eye drops) to control the progression of glaucoma and in the management of ocular hypertension.

Bimatoprost is a prostaglandin analog used topically (as eye drops) to control the progression of glaucoma and in the management of ocular hypertension.

Specifications

Chemical Name or Material Bimatoprost
Physical Form Powder
CAS 155206-00-1
Color White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C25H37NO4
Synonym AGN 192024
Solubility Information DMSO : ≥ 100 mg/mL (240.63 mM)
SMILES O=C(NCC)CCC/C=C\C[C@@H]1[C@@H](/C=C/[C@@H](O)CCC2=CC=CC=C2)[C@H](O)C[C@@H]1O
Molecular Weight (g/mol) 415.57
Formula Weight 415.57
Percent Purity 98.18%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.