missing translation for 'onlineSavingsMsg'
Learn More

Bimatoprost Acid, Mikromol

Product Code. 30461106
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30461106 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30461106 Supplier Mikromol Supplier No. MM3603.020025
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol- your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Bimatoprost Acid, Mikromol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Bimatoprost acid
CAS 38344-08-0
Analyte or Component Names Bimatoprost acid
Shipping Condition Room Temperature
Molecular Formula C23 H32 O5
Certifications/Compliance ISO 9001
Synonym 5-Heptenoic acid, 7-[(1R,2R,3R,5S)-3,5-dihydroxy-2-[(1E,3S)-3-hydroxy-5-phenyl-1-penten-1-yl]cyclopentyl]-, (5Z)-, 5-Heptenoic acid, 7-[(1R,2R,3R,5S)-3,5-dihydroxy-2-[(1E,3S)-3-hydroxy-5-phenyl-1-pentenyl]cyclopentyl]-, (5Z)- (9CI), 5-Heptenoic acid, 7-[3,5-dihydroxy-2-(3-hydroxy-5-phenyl-1-pentenyl)cyclopentyl]-, [1R-[1α(Z),2β(1E,3S*),3α,5α]]-, (5Z)-7-[(1R,2R,3R,5S)-3,5-Dihydroxy-2-[(1E,3S)-3-hydroxy-5-phenyl-1-penten-1-yl]cyclopentyl]-5-heptenoic acid, (5Z)-7-[(1R,2R,3R,5S)-3,5-Dihydroxy-2-[(S,E)-3-hydroxy-5-phenylpent-1-enyl]cyclopentyl]hept-5-enoic acid, 17-Phenyl-18,19,20-trinor-PGF2α, 17-Phenyl-18,19,20-trinorprostaglandin F2α, Bimatoprost acid, PhXA 70, U 35687
InChI Formula InChI=1S/C23H32O5/c24-18(13-12-17-8-4-3-5-9-17)14-15-20-19(21(25)16-22(20)26)10-6-1-2-7-11-23(27)28/h1,3-6,8-9,14-15,18-22,24-26H,2,7,10-13,16H2,(H,27,28)/b6-1-,15-14+/t18-,19+,20+,21-,22+/m0/s1
Solution Type Neat
SMILES O[C@@H](CCc1ccccc1)\C=C\[C@H]2[C@H](O)C[C@H](O)[C@@H]2C\C=C/CCCC(=O)O
IUPAC Name (Z)-7-[(1R,2R,3R,5S)-3,5-dihydroxy-2-[(E,3S)-3-hydroxy-5-phenylpent-1-enyl]cyclopentyl]hept-5-enoic acid
Molecular Weight (g/mol) 388.5
Quantity 25 mg
Formula Weight 388.225 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.