missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bim Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£159.00 - £376.00
Specifications
| Antigen | Bim |
|---|---|
| Concentration | 0.6 mg/mL |
| Dilution | Western Blot 2.0 ug/ml, Immunohistochemistry 1:50 - 1:100, Immunocytochemistry/Immunofluorescence 10-20 ug/ml, Immunohistochemistry-Paraffin 1:50 - 1:100 |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18109389
|
Novus Biologicals
NBP2-53088SS |
0.025 mg |
£159.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18276454
|
Novus Biologicals
NBP2-53088 |
0.1 mg |
£376.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Bim Polyclonal specifically detects Bim in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| Bim | |
| Western Blot 2.0 ug/ml, Immunohistochemistry 1:50 - 1:100, Immunocytochemistry/Immunofluorescence 10-20 ug/ml, Immunohistochemistry-Paraffin 1:50 - 1:100 | |
| Polyclonal | |
| Rabbit | |
| Apoptosis, Cancer, Immunology, Innate Immunity, Phospho Specific, Tumor Suppressors | |
| PBS with 1% BSA with 0.02% Sodium Azide | |
| BAM, bcl-2 interacting mediator of cell death, bcl-2 interacting protein Bim, Bcl2-interacting mediator of cell death, bcl2-L-11, BCL2-like 11 (apoptosis facilitator), bcl-2-like protein 11, bcl-2-related ovarian death agonist, BIM-alpha6, BIM-beta6, BIM-beta7, BIMBOD, BimEL, BimL | |
| BCL2L11 | |
| IgG | |
| Affinity Purified |
| 0.6 mg/mL | |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human, Mouse | |
| O43521 | |
| 10018 | |
| Partial recombinant human Bim protein (amino acids 2-120) UniProt O43521] | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title