missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Bim Polyclonal specifically detects Bim in Human samples. It is validated for ELISA.
Specifications
Specifications
| Antigen | Bim |
| Applications | ELISA |
| Classification | Polyclonal |
| Conjugate | DyLight 488 |
| Formulation | 50 mM Sodium Borate |
| Gene Alias | BAM, bcl-2 interacting mediator of cell death, bcl-2 interacting protein Bim, Bcl2-interacting mediator of cell death, bcl2-L-11, BCL2-like 11 (apoptosis facilitator), bcl-2-like protein 11, bcl-2-related ovarian death agonist, BIM-alpha6, BIM-beta6, BIM-beta7, BIMBOD, BimEL, BimL |
| Host Species | Rabbit |
| Immunogen | Produced in rabbits immunized with purified, recombinant Bim (NP_006529.1; Ala2-Arg120). |
| Purification Method | Affinity purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?