missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Bim Polyclonal antibody specifically detects Bim in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunofluorescence, Immunocytochemistry
Specifications
Specifications
| Antigen | Bim |
| Applications | Western Blot, ELISA, Immunofluorescence, Immunocytochemistry |
| Classification | Polyclonal |
| Conjugate | DyLight 488 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | BAM, bcl-2 interacting mediator of cell death, bcl-2 interacting protein Bim, Bcl2-interacting mediator of cell death, bcl2-L-11, BCL2-like 11 (apoptosis facilitator), bcl-2-like protein 11, bcl-2-related ovarian death agonist, BIM-alpha6, BIM-beta6, BIM-beta7, BIMBOD, BimEL, BimL |
| Host Species | Rabbit |
| Immunogen | Antibody was raised against a peptide corresponding to amino acids near the center of human Bim. The sequence is identical to that of mouse and rat. The immunogen is located within amino acids 80 - 130 of Bim. Amino Acid Squence: SGYFSFDTDRSPAPMS |
| Purification Method | Peptide affinity purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?