missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bim Antibody (1C2H4) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
£156.00
Specifications
| Antigen | Bim |
|---|---|
| Clone | 1C2H4 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 0.5-1 μg/mL, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 10 μg/mL |
| Applications | Western Blot |
Description
Bim Monoclonal specifically detects Bim in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
| Bim | |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Human | |
| BAM, bcl-2 interacting mediator of cell death, bcl-2 interacting protein Bim, Bcl2-interacting mediator of cell death, bcl2-L-11, BCL2-like 11 (apoptosis facilitator), bcl-2-like protein 11, bcl-2-related ovarian death agonist, BIM-alpha6, BIM-beta6, BIM-beta7, BIMBOD, BimEL, BimL | |
| BCL2L11 | |
| IgG1 | |
| Affinity Purified |
| 1C2H4 | |
| Western Blot 0.5-1 μg/mL, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 10 μg/mL | |
| Monoclonal | |
| Mouse | |
| Apoptosis, Cancer, Immunology, Innate Immunity, Tumor Suppressors | |
| O43521 | |
| 10018 | |
| Antibody was raised against a recombinant protein corresponding to amino acids 2 to 198 of human Bim. | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title